This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01621873_s1
See other EIF4E GE Assays ›
Gene Symbol:
Eif4e
Gene Name:
eukaryotic translation initiation factor 4E
Gene Aliases:
EG668879, Eif4e-ps, If4e, eIF-4E
Chromosome Location:
Chr.3: 138526191 - 138557599 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01621873_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007917.3 NP_031943.3 8 2332 137
GenBank mRNA AK082867.1 - 7 2213 137
BC085087.1 - 7 2204 6438419 137
M61731.1 - 7 2217 137

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4e

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
behavioral fear response
translation
translational initiation
regulation of translation
negative regulation of translation
stem cell population maintenance
negative regulation of neuron differentiation
positive regulation of mitotic cell cycle
RNA binding
translation initiation factor activity
protein binding
eukaryotic initiation factor 4G binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300011458.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011458.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011456.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011456.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011450.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011450.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011455.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011455.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011459.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011459.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011457.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011457.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011454.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011454.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011460.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011460.mm.1 TC0300001416.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300011461.mm.1 NM_007917.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300011461.mm.1 TC0300001416.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0