This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01627304_s1
See other ECHDC2 GE Assays ›
Gene Symbol:
Echdc2
Gene Name:
enoyl Coenzyme A hydratase domain containing 2
Gene Aliases:
1300017C12Rik, 2610009M20Rik, D4Ertd765e
Chromosome Location:
Chr.4: 108165436 - 108179308 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01627304_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echdc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001254754.1 NP_001241683.1 1 68 145
NM_026728.4 NP_081004.2 1 68 145
GenBank mRNA AK166388.1 - 1 40 145

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK166388.1, NM_001254754.1, NM_026728.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Echdc2

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
metabolic process
catalytic activity
lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400008039.mm.1 NM_001254754.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008039.mm.1 NM_001254754.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008039.mm.1 NM_026728.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008039.mm.1 NM_026728.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400008039.mm.1 TC0400001141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400008040.mm.1 NM_001254754.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008040.mm.1 NM_001254754.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008040.mm.1 NM_026728.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008040.mm.1 NM_026728.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400008040.mm.1 TC0400001141.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400008036.mm.1 NM_001254754.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008036.mm.1 NM_001254754.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008036.mm.1 NM_026728.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400008036.mm.1 NM_026728.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400008036.mm.1 TC0400001141.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0