This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01700423_m1
See other ISLR GE Assays ›
Gene Symbol:
Islr
Gene Name:
immunoglobulin superfamily containing leucine-rich repeat
Gene Aliases:
AW553146
Chromosome Location:
Chr.9: 58156246 - 58159239 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01700423_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Islr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012043.4 NP_036173.1 1 - 2 253 88
GenBank mRNA BC006602.1 - 1 - 2 174 3966024 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC006602.1, NM_012043.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Islr

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 NM_012043.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 AB024538.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 AI838480.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 BC006602.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 NM_001195431.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 NM_012043.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 AB024538.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 AI838480.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 BC006602.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140742 NM_001195431.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474425 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102314 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 NM_012043.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 AB024538.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 AI838480.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 BC006602.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 NM_001195431.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 NM_012043.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 AB024538.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 AI838480.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 BC006602.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140742 NM_001195431.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546121 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546143 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546149 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546135 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546164 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546154 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546139 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546136 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546173 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546123 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546130 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546138 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546129 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546159 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546165 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546125 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546171 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546133 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546147 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546128 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546140 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546151 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546124 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546122 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546170 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546145 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546131 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546148 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546157 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546162 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546163 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546120 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546156 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546152 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546161 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546169 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546167 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26546126 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102314 - -
Whole Mouse Genome G4122A A_51_P140742 NM_012043.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 AB024538.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 AI838480.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 BC006602.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 NM_001195431.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 NM_012043.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 AB024538.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 AI838480.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 BC006602.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140742 NM_001195431.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P323484 NM_012043.4 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P323484 AB024538.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P323484 BC006602.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P323484 NM_001195431.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129639 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102314 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08134376 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07573486 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07802440 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07631187 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07802438 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07978707 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08134378 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07802437 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07802439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546116 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546136 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546178 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546194 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546147 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546122 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26546119 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08000205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546164 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546140 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07573483 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08128217 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546161 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546149 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546165 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546128 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546106 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07621526 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546114 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07978703 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546119 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546138 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07573484 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546187 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08198268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07881823 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546177 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546147 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07978713 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546170 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546157 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546126 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546167 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07573486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07631187 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546120 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546183 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546151 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546178 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546136 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546155 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07978714 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546130 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546159 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546145 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08171061 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546143 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08189125 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546171 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26546113 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10594045 - -
Affymetrix Mouse Gene 1.0 ST Array 10594046 - -
Affymetrix Mouse Gene 1.0 ST Array 10594047 - -
Affymetrix HT MG-430 PM Array Plate 1418450_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10594044 - -
Mouse Genome 430 2.0 Array 1418450_PM_at - -
Mouse Genome 430 2.0 Array 1418450_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 26968_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 26968_at - -
Affymetrix Mouse Clariom S Array PSR0900020047.mm.1 NM_001195431.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020047.mm.1 NM_001195431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020047.mm.1 NM_012043.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020047.mm.1 NM_012043.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900020047.mm.1 TC0900002387.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900020050.mm.1 NM_012043.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020050.mm.1 NM_012043.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900020050.mm.1 TC0900002387.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900020049.mm.1 NM_012043.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020049.mm.1 NM_012043.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900020049.mm.1 TC0900002387.mm.1 MTA TC
Mouse Expression Set 430 1418450_PM_at - -
Mouse Expression Set 430 1418450_at - -
Murine Genome U74v2 Set 99010_at - -
Murine Genome U74v2 Set 99010_PM_at - -
Mouse Genome 430A 2.0 Array 1418450_PM_at - -
Mouse Genome 430A 2.0 Array 1418450_at - -
Affymetrix HT Mouse Genome 430A Array 1418450_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1418450_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 110273 - -
Illumina mouse-6 v1.0 expression beadchip 6450368 - -
Illumina mouse-6 v1.0 expression beadchip 105130215 - -
Illumina mouse-6 v1.0 expression beadchip 110273 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006450368 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000110273 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259262 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219747 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1219747 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1226665 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1259262 - -
mouse-6 v1.1 (Illumina) scl0003609.1_104-S - -
mouse-6 v1.1 (Illumina) scl026968.1_121-S - -
mouse-6 v1.1 (Illumina) scl0003433.1_1-S - -
Illumina Sentrix mouse-6.v1 bead array 105130215 - -
Illumina Sentrix mouse-6.v1 bead array 6450368 - -
Illumina Sentrix mouse-6.v1 bead array 110273 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0003609.1_104-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl026968.1_121-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450368 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 105130215 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 110273 - -
MouseRef-8 v2 Expression BeadChip ILMN_2613000 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613000 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226665 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl026968.1_121-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline