This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01700556_m1
See other GPR35 GE Assays ›
Gene Symbol:
Gpr35
Gene Name:
G protein-coupled receptor 35
Gene Aliases:
KPG_007
Chromosome Location:
Chr.1: 92950862 - 92986391 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01700556_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022320.4 NP_071715.3 1 - 2 399 88
GenBank mRNA AK034870.1 - 1 - 2 183 88
AK089198.1 - 1 - 2 417 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK034870.1, NM_022320.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr35

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100007775.mm.1 NM_022320.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007775.mm.1 NM_022320.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100007775.mm.1 TC0100000923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100007774.mm.1 NM_022320.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007774.mm.1 NM_022320.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100007774.mm.1 TC0100000923.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline