This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01700739_m1
See other GPR146 GE Assays ›
Gene Symbol:
Gpr146
Gene Name:
G protein-coupled receptor 146
Gene Aliases:
BC003323, PGR8
Chromosome Location:
Chr.5: 139377698 - 139396415 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01700739_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr146
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030258.4 NP_084534.2 1 - 2 81 105
GenBank mRNA AK046512.1 - 1 - 2 104 105
AK089232.1 - 1 - 2 104 105
AK154478.1 - 1 - 2 110 105
AK154708.1 - 1 - 2 102 105
AK155280.1 - 1 - 2 168 105
AK155876.1 - 1 - 2 84 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK046512.1, AK089232.1, AK154478.1, AK154708.1, AK155876.1, NM_030258.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr146

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500015445.mm.1 NM_001038703.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015445.mm.1 NM_001038703.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500015445.mm.1 NM_030258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015445.mm.1 NM_030258.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500015445.mm.1 TC0500001673.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015441.mm.1 NM_030258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015441.mm.1 NM_030258.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500015441.mm.1 TC0500001673.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015440.mm.1 NM_030258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015440.mm.1 NM_030258.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500015440.mm.1 TC0500001673.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015444.mm.1 NM_001038703.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015444.mm.1 NM_001038703.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500015444.mm.1 NM_030258.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015444.mm.1 NM_030258.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500015444.mm.1 TC0500001673.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 796107 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline