Assay Details
| Gene Symbol: Gpr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199058.1 | NP_951013.1 | 1 | 752 | 76 | |
| GenBank mRNA | AK139367.1 | - | 2 | 931 | 76 | |
| AY064488.1 | - | 1 | 752 | 76 | ||
| BC111861.1 | - | 1 | 741 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087293 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087303 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087299 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087296 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087305 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166508 | NM_199058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166508 | AK139367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166508 | AY064488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2166508 | BC111861.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144853 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166508 | NM_199058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166508 | AK139367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166508 | AY064488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2166508 | BC111861.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P487508 | NM_199058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P487508 | AY064488.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27087321 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144853 | - | - |
| Whole Mouse Genome G4122A | A_52_P487508 | NM_199058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P487508 | AY064488.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109168 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144853 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08689377 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08689418 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08689401 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08689403 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087298 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087293 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087295 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087296 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087303 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087299 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27087305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27087300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08461222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08689413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08417626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08648980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08689412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08417631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08689387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08689416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08417642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08702445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08689377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08286433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08417624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08417632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27087329 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368781 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440148_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368780 | - | - |
| Mouse Genome 430 2.0 Array | 1440148_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440148_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000014555.mm.1 | NM_199058.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014555.mm.1 | NM_199058.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014555.mm.1 | TC1000002049.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1440148_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440148_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 858933 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5720390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592996 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592996 | - | - |
| mouse-6 v1.1 (Illumina) | scl0140741.1_146-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0140741.1_146-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2592996 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592996 | - | - |

Gpr6
Genomic Map