This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01716217_g1
See other CD200R3 GE Assays ›
Gene Symbol:
Cd200r3
Gene Name:
CD200 receptor 3
Gene Aliases:
4733401I18Rik, 4833409J19Rik, AI505817, BB106877, mCD200RLb
Chromosome Location:
Chr.16: 44943596 - 45058365 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01716217_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd200r3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK046631.1 - 1 - 2 145 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB645526.1 - 1 - 2 142 Chr. 16 - 44965256 - 44966112 [+] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Cd200 receptor 2 Cd200r2 AY230198,
Cd200r1l
-49.4kb Sense Chr. 16 - 44867097 - 44915840 on NCBI Build GRCm38
Downstream coiled-coil domain containing 80 Ccdc80 2610001E17Rik,
DRO1,
Ssg1,
Urb
126.6kb Sense Chr. 16 - 45093407 - 45127924 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline