This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01720033_g1
See other 2210414F02RIK GE Assays ›
Gene Symbol:
2210414F02Rik
Gene Name:
RIKEN cDNA 2210414F02 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01720033_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2210414F02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK008923.1 - 1 - 2 200 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glucosaminyl (N-acetyl) transferase 3,
mucin type
Gcnt3 2010013H22Rik,
2210021I22Rik,
2210401J11Rik
-33.0kb Anti-sense Chr. 9 - 70031496 - 70039887 on NCBI Build GRCm38
Downstream family with sequence similarity 81,
member A
Fam81a 6430514L14Rik 15.7kb Anti-sense Chr. 9 - 70089167 - 70142555 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430170 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 392721 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline