This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01722273_g1
See other RPS24 GE Assays ›
Gene Symbol:
Rps24
Gene Name:
ribosomal protein S24
Gene Aliases:
-
Chromosome Location:
Chr.14: 24490633 - 24496146 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01722273_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011297.2 NP_035427.2 4 - 5 467 66
GenBank mRNA AK005358.1 - 4 - 5 431 66
AK012537.1 - 4 - 5 418 66
BC081457.1 - 4 - 5 413 5712639 66
X60289.1 - 4 - 5 422 66

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps24

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA processing
translation
erythrocyte homeostasis
ribosomal small subunit biogenesis
nucleotide binding
structural constituent of ribosome
translation initiation factor binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 NM_011297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 NM_011297.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 NM_207634.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 NM_207634.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 NM_207635.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 NM_207635.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001712.mm.1 TC1400000231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 NM_011297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 NM_011297.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 NM_207634.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 NM_207634.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 NM_207635.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 NM_207635.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001704.mm.1 TC1400000231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 NM_011297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 NM_011297.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 NM_207634.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 NM_207634.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 NM_207635.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 NM_207635.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001719.mm.1 TC1400000231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 NM_011297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 NM_011297.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 NM_207634.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 NM_207634.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 NM_207635.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 NM_207635.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001709.mm.1 TC1400000231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 NM_011297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 NM_011297.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 NM_207634.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 NM_207634.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 NM_207635.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 NM_207635.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001723.mm.1 TC1400000231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 NM_011297.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 NM_011297.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 NM_207634.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 NM_207634.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 NM_207635.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 NM_207635.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001710.mm.1 TC1400000231.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 902412 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline