This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01730282_gH
See other NAA20 GE Assays ›
Gene Symbol:
Naa20
Gene Name:
N(alpha)-acetyltransferase 20, NatB catalytic subunit
Gene Aliases:
1500004D14Rik, 2900026I01Rik, AU041458, D2Ertd186e, Nat5
Chromosome Location:
Chr.2: 145903206 - 145916425 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01730282_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001141965.1 NP_001135437.1 3 - 4 242 126
GenBank mRNA AK009229.1 - 3 - 4 244 126
BC009157.1 - 3 - 4 209 2648626 126
BC096451.1 - 3 - 4 196 3673241 126
BC116374.2 - 3 - 4 201 40101000 126

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

N-terminal peptidyl-methionine acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715376 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715371 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715373 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715372 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715375 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988498 BC096451.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988498 BC009157.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988498 CO802916.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988498 AK009229.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988498 AK153145.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593132 AK135150.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593132 BC003208.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593132 BC125031.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593132 BC125032.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593132 L08407.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593132 NM_007732.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341570 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690918 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681904 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596375 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79747 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718216 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826340 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56419 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161921 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112807 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988498 BC096451.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988498 BC009157.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988498 CO802916.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988498 AK009229.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988498 AK153145.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715382 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715391 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715390 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715353 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715364 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715386 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715378 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715376 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715341 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715351 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715388 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715350 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715367 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715342 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715381 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715384 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715355 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715356 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715360 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715361 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715345 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715363 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715344 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715374 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715347 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715349 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715358 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21715370 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112807 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161921 - -
Whole Mouse Genome G4122A A_51_P424828 BC096451.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424828 BC116374.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424828 BC009157.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424828 CO802916.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424828 AK009229.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139747 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112807 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161921 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01975530 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02265858 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01214209 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02265862 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715373 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715371 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715372 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715376 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21715375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715525 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715499 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715320 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715462 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21715324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715396 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715367 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01434182 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01715051 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715339 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01192820 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715351 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715348 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715411 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715382 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715353 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715394 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715363 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715361 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715358 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01430321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715379 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715398 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01715049 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01975528 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02265858 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01715057 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715388 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715376 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715407 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715355 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715350 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01430313 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715372 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715401 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715391 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715395 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715345 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715414 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715334 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715362 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01715050 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01975534 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715336 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715356 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01192824 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01290940 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02253456 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715337 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21715341 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10476776 - -
Affymetrix Mouse Gene 1.0 ST Array 10476780 - -
Affymetrix Mouse Gene 1.0 ST Array 10476779 - -
Affymetrix Mouse Gene 1.0 ST Array 10476777 - -
Affymetrix Mouse Gene 1.0 ST Array 10572933 - -
Affymetrix Mouse Gene 1.0 ST Array 10476778 - -
Affymetrix Mouse Gene 1.0 ST Array 10476781 - -
Affymetrix HT MG-430 PM Array Plate 1418244_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10572932 - -
Affymetrix Mouse Gene 1.1 ST Array 10476775 - -
Mouse Genome 430 2.0 Array 1418244_PM_at - -
Mouse Genome 430 2.0 Array 1418244_at - -
Affymetrix Mouse Clariom S Array PSR0200017135.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017135.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017135.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017135.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017135.mm.1 TC0200002170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017141.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017141.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017141.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017141.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017141.mm.1 TC0200002170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017137.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017137.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017137.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017137.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017137.mm.1 TC0200002170.mm.1 MTA TC
Mouse Expression Set 430 1418244_PM_at - -
Mouse Expression Set 430 1418244_at - -
Mouse Genome 430A 2.0 Array 1418244_PM_at - -
Mouse Genome 430A 2.0 Array 1418244_at - -
Affymetrix HT Mouse Genome 430A Array 1418244_at - -
Affymetrix HT Mouse Genome 430A Array 1418244_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4850242 - -
Illumina mouse-6 v1.0 expression beadchip 6110427 - -
Illumina mouse-6 v1.0 expression beadchip 4850242 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006110427 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004850242 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223543 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670220 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1223543 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2670220 - -
mouse-6 v1.1 (Illumina) scl067877.6_19-S - -
mouse-6 v1.1 (Illumina) scl0067877.1_158-S - -
Illumina Sentrix mouse-6.v1 bead array 4850242 - -
Illumina Sentrix mouse-6.v1 bead array 6110427 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl067877.6_19-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0067877.1_158-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850242 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110427 - -
MouseRef-8 v2 Expression BeadChip ILMN_1223543 - -
MouseRef-8 v2 Expression BeadChip ILMN_2945073 - -
MouseRef-8 v2 Expression BeadChip ILMN_2945066 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2945073 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223543 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2945066 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0