This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01743395_s1
See other RBMXL1 GE Assays ›
Gene Symbol:
Rbmxl1
Gene Name:
RNA binding motif protein, X linked-like-1
Gene Aliases:
Hnrpg, Rbmx, Rbmxrt
Chromosome Location:
Chr.8: 78505269 - 78508928 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01743395_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbmxl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252089.1 NP_001239018.1 1 77 69
NM_009033.2 NP_033059.2 1 77 69
GenBank mRNA AK149658.1 - 1 50 69
AK149764.1 - 1 50 69
BY262810.1 - 1 77 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001252089.1, NM_009033.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbmxl1

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
mRNA processing
RNA splicing
positive regulation of transcription from RNA polymerase II promoter
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
protein homooligomerization
nucleotide binding
core promoter binding
nucleic acid binding
chromatin binding
RNA binding
protein binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989165 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989164 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989167 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989163 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989168 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989169 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186621 AK207688.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186621 BC089350.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186621 AK149658.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186621 AK149764.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186621 AF031568.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186619 EF437351.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186619 BC011441.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186619 BC089350.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186619 AK088213.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186619 AK149658.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191522 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30785 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159367 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186621 AK207688.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186621 BC089350.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186621 AK149658.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186621 AK149764.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186621 AF031568.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186619 EF437351.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186619 BC011441.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186619 BC089350.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186619 AK088213.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186619 AK149658.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P447714 EF437351.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P447714 BC011441.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P447714 BC089350.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P447714 AK088213.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P447714 AK149658.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989135 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989169 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989132 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989162 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989158 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989154 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989152 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989168 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989171 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989164 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989157 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989137 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989155 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989166 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989159 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989167 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25989138 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159367 - -
Whole Mouse Genome G4122A A_51_P447714 EF437351.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P447714 BC011441.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P447714 BC089350.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P447714 AK088213.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P447714 AK149658.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129068 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159367 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07353143 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07353146 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07353138 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07353132 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07353119 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07353145 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989165 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989168 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989167 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989164 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989163 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25989169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989154 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989102 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989116 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989093 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989125 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989108 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25989169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07060635 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989158 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989124 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07435893 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989154 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989153 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989162 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989166 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989128 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989119 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06982461 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989152 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989118 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989138 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07353144 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989133 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989159 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07048399 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989167 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989131 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25989120 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29566051 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29566045 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29566050 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10580988 - -
Affymetrix Mouse Gene 1.0 ST Array 10579831 - -
Affymetrix Mouse Gene 1.0 ST Array 10580987 - -
Affymetrix Mouse Gene 1.0 ST Array 10414257 - -
Affymetrix Mouse Gene 1.0 ST Array 10579832 - -
Affymetrix HT MG-430 PM Array Plate 1416177_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10579830 - -
Affymetrix Mouse Gene 1.1 ST Array 10580986 - -
Affymetrix Mouse Gene 1.1 ST Array 10414256 - -
Mouse Genome 430 2.0 Array 1416177_PM_at - -
Mouse Genome 430 2.0 Array 1416177_at - -
Affymetrix Mouse Clariom S Array PSR0800019082.mm.1 NM_001252089.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019082.mm.1 NM_001252089.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019082.mm.1 NM_009033.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019082.mm.1 NM_009033.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019082.mm.1 TC0800002510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019084.mm.1 NM_001252089.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019084.mm.1 NM_001252089.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019084.mm.1 NM_009033.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019084.mm.1 NM_009033.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019084.mm.1 TC0800002510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019080.mm.1 NM_001252089.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019080.mm.1 NM_001252089.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019080.mm.1 NM_009033.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019080.mm.1 NM_009033.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019080.mm.1 TC0800002510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019081.mm.1 NM_009033.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019081.mm.1 NM_009033.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019081.mm.1 TC0800002510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800019083.mm.1 NM_001252089.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019083.mm.1 NM_001252089.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019083.mm.1 NM_009033.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800019083.mm.1 NM_009033.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800019083.mm.1 TC0800002510.mm.1 MTA TC
Mouse Expression Set 430 1416177_at - -
Mouse Expression Set 430 1416177_PM_at - -
Murine Genome U74v2 Set 160192_PM_at - -
Murine Genome U74v2 Set 160192_at - -
Mouse Genome 430A 2.0 Array 1416177_at - -
Mouse Genome 430A 2.0 Array 1416177_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1416177_at - -
Affymetrix HT Mouse Genome 430A Array 1416177_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2900541 - -
Illumina mouse-6 v1.0 expression beadchip 2900541 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002900541 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2765245 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2765245 - -
mouse-6 v1.1 (Illumina) scl019656.1_169-S - -
Illumina Sentrix mouse-6.v1 bead array 2900541 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl019656.1_169-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900541 - -
MouseRef-8 v2 Expression BeadChip ILMN_2765245 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2765245 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zh9xj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0