This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01964336_s1
See other FABP9 GE Assays ›
Gene Symbol:
Fabp9
Gene Name:
fatty acid binding protein 9, testis
Gene Aliases:
1700007P10Rik, Perf, Perf15, Tlbp
Chromosome Location:
Chr.3: 10193623 - 10197283 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01964336_s1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fabp9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011598.3 NP_035728.2 4 584 146
GenBank mRNA AK005745.1 - 4 528 146
AK161314.1 - 4 489 146
BC048437.1 - 4 523 6773242 146
CN837748.1 - 295 146

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fabp9

Gene Ontology Categories:

Function(s) Process(es)

transport
transporter activity
lipid binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 BC048437.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 AK161314.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 NM_011598.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 AK005745.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 BC048437.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 AK161314.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 NM_011598.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235002 AK005745.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412378 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805736 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226048 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723365 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135660 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805618 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44880 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115116 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 BC048437.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 AK161314.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 NM_011598.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 AK005745.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 BC048437.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 AK161314.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 NM_011598.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235002 AK005745.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941316 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941306 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941328 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941321 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941303 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941294 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941290 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941322 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941293 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941315 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941288 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941305 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941297 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941327 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941309 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941331 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941312 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941324 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941319 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941299 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941313 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941310 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941296 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941317 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941301 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115116 - -
Whole Mouse Genome G4122A A_51_P235002 BC048437.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 AK161314.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 NM_011598.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 AK005745.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 BC048437.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 AK161314.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 NM_011598.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P235002 AK005745.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P510518 U96149.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115116 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02735469 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02735466 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02735472 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02735465 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02735470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941317 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941306 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941228 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941350 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941288 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03078937 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941290 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941341 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941294 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941319 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941329 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941335 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941303 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03078936 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941297 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02756903 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941309 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941316 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941277 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941340 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02667497 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941313 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941347 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03155615 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941317 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02788232 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02756901 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941310 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941301 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941306 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941333 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02696589 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941328 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941343 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941322 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941305 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941283 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941327 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941296 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02891649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21941286 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10497259 - -
Affymetrix Mouse Gene 1.0 ST Array 10497263 - -
Affymetrix Mouse Gene 1.0 ST Array 10497261 - -
Affymetrix Mouse Gene 1.0 ST Array 10497262 - -
Affymetrix Mouse Gene 1.0 ST Array 10497264 - -
Affymetrix Mouse Gene 1.0 ST Array 10497260 - -
Affymetrix HT MG-430 PM Array Plate 1417076_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1421804_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10497258 - -
Mouse Genome 430 2.0 Array 1417076_PM_at - -
Mouse Genome 430 2.0 Array 1421804_PM_at - -
Mouse Genome 430 2.0 Array 1417076_at - -
Mouse Genome 430 2.0 Array 1421804_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 21884_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 21884_at - -
Affymetrix Mouse Clariom S Array PSR0300013026.mm.1 NM_011598.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300013026.mm.1 TC0300003224.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013029.mm.1 NM_011598.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300013029.mm.1 TC0300003224.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013027.mm.1 NM_011598.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300013027.mm.1 TC0300003224.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013028.mm.1 NM_011598.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300013028.mm.1 TC0300003224.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300013030.mm.1 NM_011598.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300013030.mm.1 TC0300003224.mm.1 MTA TC
Mouse Expression Set 430 1421804_at - -
Mouse Expression Set 430 1421804_PM_at - -
Mouse Expression Set 430 1417076_PM_at - -
Mouse Expression Set 430 1417076_at - -
Murine Genome U74v2 Set 160729_PM_f_at - -
Murine Genome U74v2 Set 94927_PM_at - -
Murine Genome U74v2 Set 160729_f_at - -
Murine Genome U74v2 Set 94927_at - -
Mouse Genome 430A 2.0 Array 1417076_at - -
Mouse Genome 430A 2.0 Array 1421804_at - -
Mouse Genome 430A 2.0 Array 1417076_PM_at - -
Mouse Genome 430A 2.0 Array 1421804_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1417076_at - -
Affymetrix HT Mouse Genome 430A Array 1417076_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1421804_at - -
Affymetrix HT Mouse Genome 430A Array 1421804_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 883472 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2510609 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002510609 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221705 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1221705 - -
mouse-6 v1.1 (Illumina) scl22391.4.30_3-S - -
Illumina Sentrix mouse-6.v1 bead array 2510609 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl22391.4.30_3-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510609 - -
MouseRef-8 v2 Expression BeadChip ILMN_1221705 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221705 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0