This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01966122_u1
See other EEF1A1 GE Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01966122_u1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010106.2 NP_034236.2 6 933 129
GenBank mRNA AK011771.1 - 6 882 129
AK032914.1 - 6 879 129
AK050635.1 - 6 882 130
AK076696.1 - 6 881 129
AK081725.1 - 6 881 129
AK083361.1 - 6 881 129
AK133725.1 - 6 881 129
AK135448.1 - 6 882 129
AK137223.1 - 6 881 129
AK141243.1 - 6 883 129
AK150798.1 - 6 881 129
AK151479.1 - 6 882 129
AK151773.1 - 6 881 129
AK152810.1 - 6 881 129
AK152993.1 - 6 882 129
AK153387.1 - 6 881 129
AK153402.1 - 6 881 129
AK153851.1 - 6 883 130
AK162049.1 - 6 881 129
AK162313.1 - 6 880 129
AK162440.1 - 6 885 129
AK163799.1 - 6 881 129
AK164113.1 - 6 883 129
AK164671.1 - 6 881 129
AK165938.1 - 6 881 129
AK166021.1 - 6 881 129
AK166033.1 - 6 881 129
AK166058.1 - 6 881 129
AK166089.1 - 6 881 129
AK166615.1 - 6 881 129
AK166807.1 - 6 881 129
AK167843.1 - 6 881 129
AK167873.1 - 6 880 129
AK168424.1 - 6 883 129
AK168467.1 - 6 889 129
AK190580.1 - 2 162 129
AK190942.1 - 2 162 129
AK192548.1 - 2 162 129
AK192994.1 - 2 162 129
AK194451.1 - 2 162 129
AK194947.1 - 2 162 129
AK195943.1 - 2 162 129
AK197236.1 - 2 162 129
AK197803.1 - 2 162 129
AK212450.1 - 2 162 129
AK213920.1 - 2 162 129
BC003969.1 - 5 851 129
BC040689.1 - 3 373 129
BC050124.1 - 2426 129
BC068258.1 - 2678 129
BC083069.1 - 6 869 6310448 129
BC092276.1 - 5 864 3490007 129
BC095965.1 - 5 859 6310404 129
BC108391.1 - 6 866 30242277 129
M22432.1 - 6 872 129
X13661.1 - 5 849 129

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1a1

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900022501.mm.1 NM_010106.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900022501.mm.1 NM_010106.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900022501.mm.1 TC0900002727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900022505.mm.1 NM_010106.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900022505.mm.1 NM_010106.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900022505.mm.1 TC0900002727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900022499.mm.1 NM_010106.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900022499.mm.1 NM_010106.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900022499.mm.1 TC0900002727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900022506.mm.1 NM_010106.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900022506.mm.1 NM_010106.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900022506.mm.1 TC0900002727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900022503.mm.1 NM_010106.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900022503.mm.1 NM_010106.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900022503.mm.1 TC0900002727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900022508.mm.1 NM_010106.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900022508.mm.1 NM_010106.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900022508.mm.1 TC0900002727.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0