This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02028666_g1
See other ATP5H GE Assays ›
Gene Symbol:
Atp5h
Gene Name:
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit D
Gene Aliases:
0610009D10Rik
Chromosome Location:
Chr.11: 115415691 - 115419919 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02028666_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027862.1 NP_082138.1 4 - 5 328 88
GenBank mRNA AF354051.1 - 3 - 4 321 88
AK002376.1 - 4 - 5 328 88
AK088617.1 - 3 - 4 323 88
BC081431.1 - 4 - 5 327 6408035 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5h

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
ATP synthesis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transporter activity
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100038240.mm.1 NM_027862.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038240.mm.1 NM_027862.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038240.mm.1 TC1100004071.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100038239.mm.1 NM_027862.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038239.mm.1 NM_027862.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100038239.mm.1 TC1100004071.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0