This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02342429_g1
See other PPIA GE Assays ›
Gene Symbol:
Ppia Gm17494
Gene Name:
peptidylprolyl isomerase A
predicted gene, 17494
Gene Aliases:
2700098C05, Cphn, CyP-18, CypA
-
Chromosome Location:
Chr.11: 6415870 - 6419810 on Build GRCm38
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm02342429_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008907.1 NP_032933.1 3 - 4 232 112
GenBank mRNA AK002236.1 - 3 - 4 235 112
AK007981.1 - 3 - 4 237 112
AK010649.1 - 3 - 4 235 112
AK012491.1 - 3 - 4 243 112
AK012531.1 - 3 - 4 235 112
AK012663.1 - 3 - 4 235 112
AK028210.1 - 3 - 4 235 112
AK131942.1 - 3 - 4 235 112
AK136788.1 - 3 - 4 235 112
AK151347.1 - 3 - 4 235 112
AK151583.1 - 3 - 4 324 112
AK153030.1 - 3 - 4 234 112
AK160631.1 - 3 - 4 235 112
AK160780.1 - 3 - 4 263 112
AK168035.1 - 3 - 4 235 112
AK169813.1 - 3 - 4 235 112
BC063097.1 - 2 - 3 2347 112
BC083076.1 - 3 - 4 223 6442005 112
BC087928.1 - 3 - 4 234 6533710 112
BC094632.1 - 3 - 4 215 5055863 112
BC099478.1 - 3 - 4 226 30843162 112
DQ985731.1 - 3 - 4 191 112
X52803.1 - 3 - 4 232 112
Gene Symbol: Gm17494
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK138382.1 - 2 112

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppia
Click here to find siRNAs for Gene Gm17494

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
neuron differentiation
lipid particle organization
regulation of viral genome replication
positive regulation of viral genome replication
positive regulation of protein secretion
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
isomerase activity
peptide binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100001045.mm.1 NM_008907.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001045.mm.1 NM_008907.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001045.mm.1 TC1100000096.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline