This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02342820_g1
See other EEF1G GE Assays ›
Gene Symbol:
Eef1g
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
2610301D06Rik, AA407312, EF1G
Chromosome Location:
Chr.19: 8967041 - 8978180 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02342820_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026007.4 NP_080283.3 2 - 3 344 72
GenBank mRNA AF321126.1 - 1 - 2 187 72
AK007869.1 - 2 - 3 189 72
AK007991.1 - 2 - 3 192 72
AK011951.1 - 1 - 2 186 72
AK011973.1 - 2 - 3 191 72
AK012431.1 - 2 - 3 192 72
AK014277.1 - 2 - 3 191 72
AK050636.1 - 2 - 3 195 72
AK081859.1 - 2 - 3 192 72
AK145519.1 - 2 - 3 345 72
AK145565.1 - 2 - 3 193 72
AK147980.1 - 2 - 3 192 72
AK159644.1 - 2 - 3 191 72
AK159768.1 - 2 - 3 191 72
AK166765.1 - 2 - 3 190 72
AK167831.1 - 2 - 3 192 72
AK168749.1 - 2 - 3 191 72
BC023495.1 - 1 - 2 154 5251765 72
BC083071.1 - 2 - 3 176 6329652 72
BC099413.1 - 1 - 2 172 30839877 72
BC117547.1 - 1 - 2 187 40105371 72
BC118522.1 - 1 - 2 187 40105372 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC023495.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1g

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
translation elongation factor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002322.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002322.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002322.mm.1 TC1900000170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900002328.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002328.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002328.mm.1 TC1900000170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900002329.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002329.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002329.mm.1 TC1900000170.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 381222 - -
ABI Mouse Genome Survey Microarray 893167 - -
ABI Mouse Genome Survey Microarray 896848 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0