This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02342823_g1
See other EEF1G GE Assays ›
Gene Symbol:
Eef1g
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
2610301D06Rik, AA407312, EF1G
Chromosome Location:
Chr.19: 8967041 - 8978180 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02342823_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026007.4 NP_080283.3 6 - 7 826 138
GenBank mRNA AF321126.1 - 5 - 6 669 138
AK007869.1 - 6 - 7 671 138
AK007991.1 - 6 - 7 673 138
AK011951.1 - 5 - 6 668 138
AK012431.1 - 6 - 7 672 138
AK050636.1 - 6 - 7 677 138
AK081859.1 - 6 - 7 674 138
AK145519.1 - 6 - 7 827 138
AK145565.1 - 6 - 7 675 138
AK147980.1 - 6 - 7 674 138
AK159644.1 - 6 - 7 673 138
AK159768.1 - 6 - 7 673 138
AK166765.1 - 6 - 7 672 138
AK167831.1 - 6 - 7 674 138
AK168749.1 - 6 - 7 673 138
BC023495.1 - 5 - 6 636 5251765 138
BC083071.1 - 6 - 7 658 6329652 138
BC099413.1 - 5 - 6 654 30839877 138
BC117547.1 - 5 - 6 669 40105371 138
BC118522.1 - 5 - 6 669 40105372 138

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1g

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
translation elongation factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002338.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002338.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002338.mm.1 TC1900000170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900002336.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002336.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002336.mm.1 TC1900000170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900002337.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002337.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002337.mm.1 TC1900000170.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0