This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02342825_g1
See other EEF1G GE Assays ›
Gene Symbol:
Eef1g
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
2610301D06Rik, AA407312, EF1G
Chromosome Location:
Chr.19: 8967041 - 8978180 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02342825_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026007.4 NP_080283.3 8 - 9 1203 146
GenBank mRNA AF321126.1 - 7 - 8 146
AK007869.1 - 8 - 9 1048 146
AK007991.1 - 8 - 9 1051 146
AK011951.1 - 7 - 8 1045 146
AK012431.1 - 8 - 9 1049 146
AK050636.1 - 8 - 9 1054 146
AK081859.1 - 8 - 9 1051 146
AK145519.1 - 8 - 9 1204 146
AK145565.1 - 8 - 9 1052 146
AK147980.1 - 8 - 9 1051 146
AK159644.1 - 8 - 9 1049 146
AK159768.1 - 8 - 9 1049 146
AK166765.1 - 8 - 9 1049 146
AK167831.1 - 8 - 9 1051 146
AK168749.1 - 8 - 9 1050 146
BC023495.1 - 7 - 8 1013 5251765 146
BC083071.1 - 8 - 9 1035 6329652 146
BC099413.1 - 7 - 8 1031 30839877 146
BC117547.1 - 7 - 8 1046 40105371 146
BC118522.1 - 7 - 8 1046 40105372 146

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1g

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
translation elongation factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002340.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002340.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002340.mm.1 TC1900000170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900002339.mm.1 NM_026007.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900002339.mm.1 NM_026007.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900002339.mm.1 TC1900000170.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0