This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02344177_m1
See other DBX1 GE Assays ›
Gene Symbol:
Dbx1
Gene Name:
developing brain homeobox 1
Gene Aliases:
AI426026, Dbx
Chromosome Location:
Chr.7: 49631499 - 49636835 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02344177_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dbx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005232.1 NP_001005232.1 1 - 2 483 133
GenBank mRNA BC082541.1 - 1 - 2 483 5718470 133
S75837.1 - 1 - 2 496 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003079.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dbx1

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
multicellular organism development
ventral spinal cord interneuron specification
DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700028073.mm.1 NM_001005232.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700028073.mm.1 NM_001005232.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700028073.mm.1 TC0700003079.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700028072.mm.1 NM_001005232.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700028072.mm.1 NM_001005232.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700028072.mm.1 TC0700003079.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline