This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02580988_m1
See other UBE3A GE Assays ›
Gene Symbol:
Ube3a
Gene Name:
ubiquitin protein ligase E3A
Gene Aliases:
4732496B02, 5830462N02Rik, A130086L21Rik, Hpve6a, mKIAA4216
Chromosome Location:
Chr.7: 59227814 - 59311536 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02580988_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033962.1 NP_001029134.1 4 - 5 2100 102
NM_011668.2 NP_035798.2 6 - 7 2287 102
NM_173010.3 NP_766598.1 4 - 5 2102 102
GenBank mRNA AK029133.1 - 4 - 5 2103 102
AK083795.1 - 4 - 5 2032 102
AK220549.1 - 4 - 5 2000 102
BC131658.1 - 4 - 5 2088 40110636 102
BC131659.1 - 4 - 5 2088 40110637 102
U82122.1 - 2 - 3 1537 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000949.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube3a

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
ubiquitin-dependent protein catabolic process
positive regulation of phosphatidylinositol 3-kinase signaling
protein ubiquitination
androgen receptor signaling pathway
positive regulation of protein ubiquitination
sperm entry
regulation of circadian rhythm
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
protein autoubiquitination
prostate gland growth
protein K48-linked ubiquitination
regulation of nucleic acid-templated transcription
regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
transcription coactivator activity
ubiquitin-protein transferase activity
protein binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 TC0700000949.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 NM_001033962.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 NM_001033962.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 NM_011668.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 NM_011668.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 NM_173010.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008720.mm.1 NM_173010.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 TC0700000949.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 NM_001033962.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 NM_001033962.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 NM_011668.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 NM_011668.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 NM_173010.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008724.mm.1 NM_173010.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0