This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02600127_g1
See other NDUFS5 GE Assays ›
Gene Symbol:
Ndufs5 BC002163
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 5
NADH dehydrogenase Fe-S protein 5 pseudogene
Gene Aliases:
AI256693
-
Chromosome Location:
Chr.4: 123712710 - 123718186 on Build GRCm38
Chromosome Location:
Chr.16: 42955580 - 42956079 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm02600127_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufs5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030274.1 NP_001025445.1 2 - 3 290 67
GenBank mRNA AK148505.1 - 2 - 3 289 67
AK152845.1 - 2 - 3 285 67
BC081434.1 - 2 - 3 234 6412214 67
BU534760.1 - 2 - 3 293 67
Gene Symbol: BC002163
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002445.2 1 265 67
GenBank mRNA BC002163.1 - 1 277 67

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufs5
Click here to find siRNAs for Gene BC002163

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

transport
oxidation-reduction process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23264438 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23264437 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23264435 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029750 AK148505.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029750 AK152845.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029750 BC081434.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029750 NM_001030274.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029750 NR_002445.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262694 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918450 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482574 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135026 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029750 AK148505.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029750 AK152845.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029750 BC081434.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029750 NM_001030274.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029750 NR_002445.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264427 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264422 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264439 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264452 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264431 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264437 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264430 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264435 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264425 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264441 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264450 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264453 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264454 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23264445 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05385207 - -
Whole Mouse Genome G4122A A_51_P214916 BU534760.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P214916 AK148505.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P214916 AK152845.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P214916 BC081434.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P214916 NM_001030274.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05385207 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03755097 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03755098 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23264437 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23264435 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23264438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264417 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23264448 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03251368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264428 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03364697 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264427 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264420 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264431 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264441 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03409942 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264435 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03475828 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264437 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264412 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264452 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264443 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03364695 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03364696 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264404 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264440 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264445 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264424 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264439 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264411 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264397 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264414 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03967415 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264438 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264417 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264422 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23264433 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712375 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13048663 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712372 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712362 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712370 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712383 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13048678 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712387 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712394 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712376 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712388 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712360 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712366 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712385 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712382 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13023548 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712402 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712397 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712367 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712384 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12485294 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712364 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712381 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712400 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712391 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712390 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30712396 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10435785 - -
Affymetrix Mouse Gene 1.0 ST Array 10516212 - -
Affymetrix Mouse Gene 1.0 ST Array 10435786 - -
Affymetrix Mouse Gene 1.0 ST Array 10435786 - -
Affymetrix Mouse Gene 1.0 ST Array 10435785 - -
Affymetrix HT MG-430 PM Array Plate 1416494_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1416495_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10435784 - -
Affymetrix Mouse Gene 1.1 ST Array 10516211 - -
Affymetrix Mouse Gene 1.1 ST Array 10435784 - -
Mouse Genome 430 2.0 Array 1416494_PM_at - -
Mouse Genome 430 2.0 Array 1416495_PM_s_at - -
Mouse Genome 430 2.0 Array 1416495_s_at - -
Mouse Genome 430 2.0 Array 1416494_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 595136_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 595136_PM_at - -
Affymetrix Mouse Clariom S Array PSR0400028351.mm.1 TC0400003489.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028351.mm.1 NM_001030274.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028351.mm.1 NM_001030274.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028352.mm.1 TC0400003489.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028352.mm.1 NM_001030274.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028352.mm.1 NM_001030274.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028358.mm.1 TC0400003489.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028358.mm.1 NM_001030274.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028358.mm.1 NM_001030274.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005559.mm.1 NR_002445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600005559.mm.1 TC1600000620.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005560.mm.1 NR_002445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600005560.mm.1 TC1600000620.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005561.mm.1 NR_002445.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600005561.mm.1 TC1600000620.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028350.mm.1 TC0400003489.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028350.mm.1 NM_001030274.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028350.mm.1 NM_001030274.1 Refseq
Mouse Expression Set 430 1416494_at - -
Mouse Expression Set 430 1416495_s_at - -
Mouse Expression Set 430 1416494_PM_at - -
Mouse Expression Set 430 1416494_PM_at - -
Mouse Expression Set 430 1416495_PM_s_at - -
Mouse Expression Set 430 1416495_s_at - -
Mouse Expression Set 430 1416494_at - -
Mouse Expression Set 430 1416495_PM_s_at - -
Murine Genome U74v2 Set 99593_at - -
Murine Genome U74v2 Set 99593_at - -
Murine Genome U74v2 Set 99593_PM_at - -
Murine Genome U74v2 Set 99593_PM_at - -
Mouse Genome 430A 2.0 Array 1416495_s_at - -
Mouse Genome 430A 2.0 Array 1416494_PM_at - -
Mouse Genome 430A 2.0 Array 1416495_s_at - -
Mouse Genome 430A 2.0 Array 1416495_PM_s_at - -
Mouse Genome 430A 2.0 Array 1416494_at - -
Mouse Genome 430A 2.0 Array 1416494_at - -
Mouse Genome 430A 2.0 Array 1416494_PM_at - -
Mouse Genome 430A 2.0 Array 1416495_PM_s_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 898986 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259050 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1259050 - -
MouseRef-8 v2 Expression BeadChip ILMN_2946817 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946817 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0