This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02600478_g1
See other IGSF9B GE Assays ›
Gene Symbol:
Igsf9b
Gene Name:
immunoglobulin superfamily, member 9B
Gene Aliases:
Gm508, mKIAA1030
Chromosome Location:
Chr.9: 27299104 - 27357549 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02600478_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf9b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033323.3 NP_001028495.2 10 - 11 1599 63
NM_001129787.1 NP_001123259.1 10 - 11 1599 63
GenBank mRNA AK158950.1 - 10 - 11 1599 63
BC150882.1 - 10 - 11 1407 63
BC150887.1 - 10 - 11 1407 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000259.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
positive regulation of inhibitory postsynaptic potential
kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001955.mm.1 NM_001129787.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001955.mm.1 NM_001129787.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001955.mm.1 TC0900000259.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001956.mm.1 NM_001129787.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001956.mm.1 NM_001129787.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001956.mm.1 TC0900000259.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0