Assay Details
| Gene Symbol: Rpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009081.2 | NP_033107.1 | 4 - 5 | 368 | 78 | |
| GenBank mRNA | AK012368.1 | - | 4 - 5 | 369 | 78 | |
| AK018714.1 | - | 4 - 5 | 369 | 78 | ||
| AK076133.1 | - | 4 - 5 | 369 | 78 | ||
| BC024395.1 | - | 4 - 5 | 368 | 4981342 | 78 | |
| BC086797.1 | - | 4 - 5 | 354 | 5713507 | 78 | |
| BC086932.1 | - | 4 - 5 | 370 | 6760499 | 78 | |
| BC092012.1 | - | 4 - 5 | 368 | 30608657 | 78 | |
| X74856.1 | - | 4 - 5 | 396 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950545 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950543 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950546 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985359 | AK012368.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985359 | NM_009081.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985359 | AK018714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985359 | AK076133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985359 | BC024395.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666338 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132151 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044109 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985359 | AK012368.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985359 | NM_009081.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985359 | AK018714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985359 | AK076133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985359 | BC024395.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339103 | AK012368.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339103 | NM_009081.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339103 | AK018714.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339103 | AK076133.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P339103 | BC024395.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132151 | - | - |
| Whole Mouse Genome G4122A | A_51_P339103 | AK012368.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339103 | NM_009081.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339103 | AK018714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339103 | AK076133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P339103 | BC024395.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104479 | AK076133.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104479 | BC086797.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104479 | X74856.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132151 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950543 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950547 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950546 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24950545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06011210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06584345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06584341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06011206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06630849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06371152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06318549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06204867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06584344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06312760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06584342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06063028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06320951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06204866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06320952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06371154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06320947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06605564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950563 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10349479 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549718 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10349478 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516275 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526179 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518329 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549714 | - | - |
| Mouse Genome 430 2.0 Array | 1416074_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416074_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000569.mm.1 | TC0700000044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000569.mm.1 | NM_009081.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000569.mm.1 | NM_009081.2 | Refseq |
| Mouse Expression Set 430 | 1416074_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416074_a_at | - | - |
| Murine Genome U74v2 Set | 100727_PM_at | - | - |
| Murine Genome U74v2 Set | 100727_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416074_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416074_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6900136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100730528 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259243 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259243 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2453804 | - | - |
| mouse-6 v1.1 (Illumina) | scl33127.5.1_128-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0019943.2_84-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100730528 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0019943.2_84-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900136 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100730528 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2759235 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2759235 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2453804 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0019943.2_84-S | - | - |

Rpl28
Genomic Map