This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02745712_m1
See other SPINT5 GE Assays ›
Gene Symbol:
Spint5
Gene Name:
serine protease inhibitor, Kunitz type 5
Gene Aliases:
Gm14316, OTTMUSG00000016293
Chromosome Location:
Chr.2: 164715305 - 164718068 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02745712_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spint5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040055.1 NP_001035144.1 3 - 4 350 71
GenBank mRNA AY542490.1 - 3 - 4 350 71
BC147380.1 - 3 - 4 332 9056450 71
BC147381.1 - 3 - 4 332 9056470 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002529.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973264 BC147380.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973264 BC147381.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973264 AY542490.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1973264 NM_001040055.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871457 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939201 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123100 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973264 BC147380.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973264 BC147381.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973264 AY542490.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1973264 NM_001040055.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04772798 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04772798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819322 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819336 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01592112 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819309 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819372 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819333 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819376 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819311 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819378 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819322 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819327 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819357 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819334 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819374 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01810332 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02183162 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819358 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01592100 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819325 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819373 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819348 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819368 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01592102 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819356 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01838517 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819341 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819375 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01499546 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01466542 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819369 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819350 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819343 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819330 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819338 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819367 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819365 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819380 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01838519 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21819360 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02132105 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10478555 - -
Affymetrix Mouse Gene 1.0 ST Array 10478556 - -
Affymetrix Mouse Gene 1.0 ST Array 10478559 - -
Affymetrix Mouse Gene 1.0 ST Array 10478557 - -
Affymetrix Mouse Gene 1.1 ST Array 10478554 - -
Affymetrix Mouse Clariom S Array PSR0200020652.mm.1 NM_001040055.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020652.mm.1 NM_001040055.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020652.mm.1 TC0200002529.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200020658.mm.1 NM_001040055.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020658.mm.1 NM_001040055.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020658.mm.1 TC0200002529.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200020656.mm.1 NM_001040055.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020656.mm.1 NM_001040055.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020656.mm.1 TC0200002529.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200020657.mm.1 NM_001040055.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020657.mm.1 NM_001040055.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200020657.mm.1 TC0200002529.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
MouseRef-8 v2 Expression BeadChip ILMN_2858548 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2858548 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline