This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03012887_g1
See other TUFM GE Assays ›
Gene Symbol:
Tufm
Gene Name:
Tu translation elongation factor, mitochondrial
Gene Aliases:
2300002G02Rik, C76308, C76389, EF-TuMT, EFTU
Chromosome Location:
Chr.7: 126487265 - 126490731 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03012887_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tufm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163713.1 NP_001157185.1 1 - 2 139 78
NM_172745.3 NP_766333.1 1 - 2 139 78
GenBank mRNA AK075857.1 - 1 - 2 63 78
AK084724.1 - 1 - 2 120 78
AK152858.1 - 1 - 2 134 78
AK153135.1 - 1 - 2 134 78
BC060959.1 - 1 - 2 116 30379757 78
BC100596.1 - 1 - 2 132 6762419 78
BY246546.1 - 1 - 2 139 78

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tufm

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
nucleotide binding
translation elongation factor activity
GTPase activity
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700015374.mm.1 TC0700001804.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700015374.mm.1 NM_001163713.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015374.mm.1 NM_001163713.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015374.mm.1 NM_172745.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015374.mm.1 NM_172745.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700015375.mm.1 TC0700001804.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700015375.mm.1 NM_001163713.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015375.mm.1 NM_001163713.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015375.mm.1 NM_172745.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700015375.mm.1 NM_172745.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0