This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03014848_m1
See other AMER3 GE Assays ›
Gene Symbol:
Amer3
Gene Name:
APC membrane recruitment 3
Gene Aliases:
9430069J07Rik, Fam123c
Chromosome Location:
Chr.1: 34578935 - 34590948 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03014848_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Amer3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213727.2 NP_998892.1 1 - 2 75 62
GenBank mRNA BC070435.1 - 1 - 2 41 6853754 62
BY290690.1 - 1 - 2 76 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC070435.1, NM_213727.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Amer3

Gene Ontology Categories:

Function(s) Process(es)

biological_process
Wnt signaling pathway
molecular_function
lipid binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20139528 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20139531 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20139530 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 NM_213727.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 AK034979.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 AW049092.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 BC070435.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 NM_213727.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 AK034979.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 AW049092.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P255304 BC070435.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954882 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855813 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885525 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235556 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506598 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520609 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42147 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117687 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110754 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 NM_213727.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 AK034979.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 AW049092.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 BC070435.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 NM_213727.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 AK034979.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 AW049092.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P255304 BC070435.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139516 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139509 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139528 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139542 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139519 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139540 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139530 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139521 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139535 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139511 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139526 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139536 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139505 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139544 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139504 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139518 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139539 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139529 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139524 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139538 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139533 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139522 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139543 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139541 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139531 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139514 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139513 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139525 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20139507 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117687 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110754 - -
Whole Mouse Genome G4122A A_52_P255304 NM_213727.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 AK034979.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 AW049092.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 BC070435.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 NM_213727.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 AK034979.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 AW049092.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P255304 BC070435.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117458 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117687 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110754 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250164 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250185 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250175 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250174 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00250176 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20139531 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20139528 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20139530 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139565 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139530 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139556 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139557 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139541 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139580 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20139552 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139642 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139627 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807308 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139629 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00072624 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00921827 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00607007 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00981883 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00417665 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139622 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139643 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139647 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139514 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250164 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139630 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250177 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139648 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139500 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250176 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139542 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250163 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139551 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250166 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00404954 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139513 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139498 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250172 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00417670 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00569022 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250182 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139539 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139638 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250178 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139515 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139634 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250175 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807319 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00800903 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00404953 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139516 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139552 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00417672 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00607000 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139535 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250188 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139548 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807331 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139625 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139624 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01100125 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139580 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807323 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00687754 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00404951 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00924725 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139639 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139644 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139655 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00250185 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00924723 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139554 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807311 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139571 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139525 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139556 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00606998 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139505 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00807320 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139510 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139526 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01100128 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139502 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20139511 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10345402 - -
Affymetrix Mouse Gene 1.0 ST Array 10345403 - -
Affymetrix HT MG-430 PM Array Plate 1436407_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10345401 - -
Mouse Genome 430 2.0 Array 1436407_PM_at - -
Mouse Genome 430 2.0 Array 1436407_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 211383_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 211383_at - -
Affymetrix Mouse Clariom S Array PSR0100001476.mm.1 NM_213727.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001476.mm.1 NM_213727.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100001476.mm.1 TC0100000228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001477.mm.1 NM_213727.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001477.mm.1 NM_213727.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100001477.mm.1 TC0100000228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001478.mm.1 NM_213727.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001478.mm.1 NM_213727.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100001478.mm.1 TC0100000228.mm.1 MTA TC
Mouse Expression Set 430 1436407_at - -
Mouse Expression Set 430 1436407_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1436407_at - -
Affymetrix HT Mouse Genome 430B Array 1436407_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 105340017 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2698061 - -
mouse-6 v1.1 (Illumina) scl18087.2_490-S - -
Illumina Sentrix mouse-6.v1 bead array 105340017 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 105340017 - -
MouseRef-8 v2 Expression BeadChip ILMN_2856946 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2698061 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2856946 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0