This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03015238_g1
See other PMP2 GE Assays ›
Gene Symbol:
Pmp2
Gene Name:
peripheral myelin protein 2
Gene Aliases:
P2
Chromosome Location:
Chr.3: 10179851 - 10183885 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03015238_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pmp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030305.2 NP_001025476.1 2 - 3 310 86
GenBank mRNA BC099520.1 - 2 - 3 310 30848207 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003223.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pmp2

Gene Ontology Categories:

Function(s) Process(es)

transport
membrane organization
transporter activity
fatty acid binding
lipid binding
cholesterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300013021.mm.1 NM_001030305.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300013021.mm.1 NM_001030305.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300013021.mm.1 TC0300003223.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline