This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03015314_g1
See other GTF3C3 GE Assays ›
Gene Symbol:
Gtf3c3
Gene Name:
general transcription factor IIIC, polypeptide 3
Gene Aliases:
AL022818, TF3C-gamma, TFIIIC102
Chromosome Location:
Chr.1: 54397581 - 54438971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03015314_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033194.2 NP_001028366.1 1 - 2 157 83
GenBank mRNA AK145040.1 - 158 83
AK160997.1 - 1 - 2 145 83
AK165830.1 - 1 - 2 158 83
BC141157.1 - 1 - 2 147 9055769 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002348.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf3c3

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100018984.mm.1 NM_001033194.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100018984.mm.1 TC0100002348.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100018985.mm.1 NM_001033194.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100018985.mm.1 TC0100002348.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0