Assay Details
| Gene Symbol: Serpinb6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045535.2 | NP_001039000.2 | 5 - 6 | 683 | 89 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965363 | NM_001045535.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520565 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63824 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965363 | NM_001045535.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28935322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10874579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10678605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10917815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10495236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10476254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10678601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28935341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408592 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408587 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408588 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441918_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408585 | - | - |
| Mouse Genome 430 2.0 Array | 1441918_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1441918_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 435350_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 435350_PM_at | - | - |
| Mouse Expression Set 430 | 1441918_x_at | - | - |
| Mouse Expression Set 430 | 1441918_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441918_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441918_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2803967 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803967 | - | - |

Serpinb6e
Genomic Map