This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03032049_gH
See other HIST1H4B GE Assays ›
Gene Symbol:
Hist1h4b 4930558J22Rik
Gene Name:
histone cluster 1, H4b
RIKEN cDNA 4930558J22 gene
Gene Aliases:
-
-
Chromosome Location:
Chr.13: 23756937 - 23757386 on Build GRCm38
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm03032049_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hist1h4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178193.2 NP_835500.1 1 155 144
GenBank mRNA BC132186.1 - 1 155 40130463 144
BC132212.1 - 1 155 40130489 144
Gene Symbol: 4930558J22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK133404.1 - 2 144

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK133404.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
DNA replication-dependent nucleosome assembly
DNA replication-independent nucleosome assembly
negative regulation of megakaryocyte differentiation
protein heterotetramerization
histone binding
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883302 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883338 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883305 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883342 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883315 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883323 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883309 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883332 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883333 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883340 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883325 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883348 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883346 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883330 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883317 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883319 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883304 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883349 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883313 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883310 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883318 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883327 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883345 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28883321 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10925254 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10925270 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10925271 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10925276 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883335 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883302 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28883338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883332 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883317 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883280 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28883270 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883318 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883349 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883332 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10703464 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883338 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883327 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883333 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883321 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883330 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883315 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883335 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883342 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883317 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10604440 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883302 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883340 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883310 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883345 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883348 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883329 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883304 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883319 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883305 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883325 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28883323 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10408093 - -
Affymetrix Mouse Gene 1.0 ST Array 10404068 - -
Affymetrix Mouse Gene 1.0 ST Array 10408075 - -
Affymetrix Mouse Gene 1.0 ST Array 10403958 - -
Affymetrix Mouse Gene 1.0 ST Array 10408244 - -
Affymetrix Mouse Gene 1.1 ST Array 10403957 - -
Affymetrix Mouse Gene 1.1 ST Array 10404067 - -
Affymetrix Mouse Gene 1.1 ST Array 10408243 - -
Affymetrix Mouse Gene 1.1 ST Array 10408092 - -
Affymetrix Mouse Gene 1.1 ST Array 10408074 - -
Mouse Genome 430 2.0 Array 1431615_PM_at - -
Mouse Genome 430 2.0 Array 1431615_at - -
Affymetrix Mouse Clariom S Array PSR1300001455.mm.1 TC1300000261.mm.1 MTA TC
Mouse Expression Set 430 1422947_at - -
Mouse Expression Set 430 1428014_PM_at - -
Mouse Expression Set 430 1422948_s_at - -
Mouse Expression Set 430 1428014_at - -
Mouse Expression Set 430 1424854_at - -
Mouse Expression Set 430 1422948_PM_s_at - -
Mouse Expression Set 430 1422947_PM_at - -
Mouse Expression Set 430 1424854_PM_at - -
Mouse Expression Set 430 1431658_at - -
Mouse Expression Set 430 1431658_PM_at - -
Mouse Expression Set 430 1431615_PM_at - -
Mouse Expression Set 430 1431615_at - -
Mouse Genome 430A 2.0 Array 1424854_PM_at - -
Mouse Genome 430A 2.0 Array 1422947_PM_at - -
Mouse Genome 430A 2.0 Array 1428014_at - -
Mouse Genome 430A 2.0 Array 1424854_at - -
Mouse Genome 430A 2.0 Array 1428014_PM_at - -
Mouse Genome 430A 2.0 Array 1422947_at - -
Mouse Genome 430A 2.0 Array 1422948_s_at - -
Mouse Genome 430A 2.0 Array 1422948_PM_s_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 818147 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4210524 - -
Illumina mouse-6 v1.0 expression beadchip 107040167 - -
Illumina mouse-6 v1.0 expression beadchip 102370338 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004210524 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213257 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1221724 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1213257 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2525620 - -
mouse-6 v1.1 (Illumina) scl44205.2.1_2-S - -
mouse-6 v1.1 (Illumina) GI_30061404-S - -
mouse-6 v1.1 (Illumina) scl00326620.1_9-S - -
Illumina Sentrix mouse-6.v1 bead array 102370338 - -
Illumina Sentrix mouse-6.v1 bead array 107040167 - -
Illumina Sentrix mouse-6.v1 bead array 4210524 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl00326620.1_9-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102370338 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210524 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 107040167 - -
MouseRef-8 v2 Expression BeadChip ILMN_1213257 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213257 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221724 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2525620 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0