This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03032054_gH
See other HIST1H2BF GE Assays ›
Gene Symbol:
Hist1h2bf Hist1h2bc Hist1h2be Hist1h2bg Hist1h2bh Hist1h2bj Hist1h2bk Hist1h2bl Hist1h2bn Hist1h2bp Hist1h2bq Hist1h2br
Gene Name:
histone cluster 1, H2bf
histone cluster 1, H2bc
histone cluster 1, H2be
histone cluster 1, H2bg
histone cluster 1, H2bh
histone cluster 1, H2bj
histone cluster 1, H2bk
histone cluster 1, H2bl
histone cluster 1, H2bn
histone cluster 1, H2bp
histone cluster 1, H2bq
histone cluster 1 H2br
Gene Aliases:
-
2610022J01Rik, H2bfs, R74621
-
-
H2b-221
-
-
H2b-291a
-
-
Gm13646, OTTMUSG00000013203
Gm11277, OTTMUSG00000000449
Chromosome Location:
Chr.13: 23573760 - 23574190 on Build GRCm38
Chromosome Location:
Chr.13: 23684199 - 23692480 on Build GRCm38
Chromosome Location:
Chr.13: 23583670 - 23621124 on Build GRCm38
Chromosome Location:
Chr.13: 23571400 - 23571863 on Build GRCm38
Chromosome Location:
Chr.13: 23542923 - 23543444 on Build GRCm38
Chromosome Location:
Chr.13: 22043230 - 22043658 on Build GRCm38
Chromosome Location:
Chr.13: 22035821 - 22036320 on Build GRCm38
Chromosome Location:
Chr.13: 21715713 - 21716143 on Build GRCm38
Chromosome Location:
Chr.13: 21754123 - 21754553 on Build GRCm38
Chromosome Location:
Chr.13: 21787480 - 21801561 on Build GRCm38
Chromosome Location:
Chr.13: 21806412 - 21810199 on Build GRCm38
Chromosome Location:
Chr.13: 21833743 - 21837530 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
-
-
-
-
-
Assay ID Mm03032054_gH
Size
Availability Inventoried
Catalog # 4331182
Price 272.00
Your Price
Online offer: 256.65
256.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hist1h2bf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178195.2 NP_835502.1 1 135 121
Gene Symbol: Hist1h2bc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290380.1 NP_001277309.1 1 168 121
NM_023422.3 NP_075911.2 1 168 121
GenBank mRNA AK005191.1 - 1 164 121
AK005407.1 - 1 163 121
AK011516.1 - 1 170 121
BC019673.1 - 1 151 3993954 121
Gene Symbol: Hist1h2be
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177653.1 NP_001171124.1 2 328 121
NM_001290530.1 NP_001277459.1 1 149 121
NM_178194.4 NP_835501.1 2 354 121
GenBank mRNA AK030546.1 - 2 356 121
AK049948.1 - 2 329 121
BC069889.1 - 1 142 4217814 121
Gene Symbol: Hist1h2bg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178196.4 NP_835503.1 1 168 121
GenBank mRNA BC060304.1 - 1 195 4217587 121
Gene Symbol: Hist1h2bh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178197.2 NP_835504.1 1 227 121
GenBank mRNA AK158416.1 - 176 121
BC092138.1 - 142 30613720 121
BC132584.1 - 1 227 40130861 121
BC132586.1 - 1 227 40130863 121
Gene Symbol: Hist1h2bj
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178198.2 NP_835505.1 1 121
Gene Symbol: Hist1h2bk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175665.2 NP_783596.1 1 201 121
GenBank mRNA BC147143.1 - 1 201 9056184 121
BC147144.1 - 1 201 9056197 121
Gene Symbol: Hist1h2bl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178199.2 NP_835506.1 1 135 121
Gene Symbol: Hist1h2bn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178201.2 NP_835508.1 1 135 121
Gene Symbol: Hist1h2bp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290466.1 NP_001277395.1 1 162 121
NM_178202.2 NP_835509.2 1 162 121
GenBank mRNA BC061044.1 - 1 162 6742370 121
Gene Symbol: Hist1h2bq
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097979.2 NP_001091448.2 1 163 121
Gene Symbol: Hist1h2br
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110555.1 NP_001104025.1 1 163 121
GenBank mRNA AK036389.1 - 1 164 121
BC011440.1 - 1 148 3989862 121

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hist1h2bf
Click here to find siRNAs for Gene Hist1h2bc
Click here to find siRNAs for Gene Hist1h2be
Click here to find siRNAs for Gene Hist1h2bg
Click here to find siRNAs for Gene Hist1h2bh
Click here to find siRNAs for Gene Hist1h2bj
Click here to find siRNAs for Gene Hist1h2bk
Click here to find siRNAs for Gene Hist1h2bl
Click here to find siRNAs for Gene Hist1h2bn
Click here to find siRNAs for Gene Hist1h2bp
Click here to find siRNAs for Gene Hist1h2bq
Click here to find siRNAs for Gene Hist1h2br

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

innate immune response in mucosa
nucleosome assembly
antibacterial humoral response
defense response to Gram-positive bacterium
biological_process
molecular_function
DNA binding
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874477 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874460 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874463 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874445 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874444 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874442 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874462 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874472 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874470 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874468 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874446 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874455 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874464 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874466 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874452 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874465 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874459 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874456 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28874475 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10893023 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10893030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874481 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874483 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874477 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28874482 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874481 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874447 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874459 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874456 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874452 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874467 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874475 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874445 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874436 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874466 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874463 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874473 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10893023 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874455 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874465 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874468 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874479 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874470 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874446 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874449 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28874440 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10940142 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10408211 - -
Affymetrix Mouse Gene 1.0 ST Array 10403979 - -
Affymetrix Mouse Gene 1.0 ST Array 10408091 - -
Affymetrix Mouse Gene 1.0 ST Array 10408071 - -
Affymetrix Mouse Gene 1.0 ST Array 10403960 - -
Affymetrix Mouse Gene 1.0 ST Array 10403981 - -
Affymetrix Mouse Gene 1.0 ST Array 10403949 - -
Affymetrix Mouse Gene 1.1 ST Array 10408070 - -
Affymetrix Mouse Gene 1.1 ST Array 10403980 - -
Affymetrix Mouse Gene 1.1 ST Array 10403959 - -
Affymetrix Mouse Gene 1.1 ST Array 10403948 - -
Affymetrix Mouse Gene 1.1 ST Array 10408210 - -
Affymetrix Mouse Gene 1.1 ST Array 10408087 - -
Affymetrix Mouse Gene 1.1 ST Array 10403978 - -
Affymetrix Mouse Clariom S Array PSR1300010117.mm.1 TC1300001630.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300001182.mm.1 NM_178201.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300001182.mm.1 TC1300000177.mm.1 MTA TC
Mouse Expression Set 430 1425398_at - -
Mouse Expression Set 430 1425398_PM_at - -
Mouse Expression Set 430 1452540_PM_a_at - -
Mouse Expression Set 430 1452540_a_at - -
Mouse Genome 430A 2.0 Array 1425398_at - -
Mouse Genome 430A 2.0 Array 1452540_a_at - -
Mouse Genome 430A 2.0 Array 1452540_PM_a_at - -
Mouse Genome 430A 2.0 Array 1425398_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 730128 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000730128 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257520 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1257520 - -
mouse-6 v1.1 (Illumina) scl0319187.1_0-S - -
Illumina Sentrix mouse-6.v1 bead array 730128 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0319187.1_0-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 730128 - -
MouseRef-8 v2 Expression BeadChip ILMN_1257520 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257520 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0319187.1_0-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline