This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03039402_g1
See other RBPJ GE Assays ›
Gene Symbol:
Rbpj
Gene Name:
recombination signal binding protein for immunoglobulin kappa J region
Gene Aliases:
AI843960, CBF1, Igkjrb, Igkrsbp, RBP-J, RBP-J kappa, RBPjk, Rbpsuh
Chromosome Location:
Chr.5: 53466117 - 53657445 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03039402_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpj
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080927.2 NP_001074396.1 9 - 10 1267 68
NM_001080928.1 NP_001074397.1 9 - 10 1153 68
NM_001277116.1 NP_001264045.1 11 - 12 1349 68
NM_009035.5 NP_033061.3 9 - 10 1169 68
GenBank mRNA AK028363.1 - 11 - 12 1327 68
AK090006.1 - 9 - 10 1054 68
AK132145.1 - 9 - 10 1109 68
AK139087.1 - 9 - 10 1054 68
AK145187.1 - 9 - 10 1061 68
AK152746.1 - 9 - 10 1169 68
AK153163.1 - 10 - 11 1057 68
BC035299.1 - 7 - 8 955 68
BC051387.1 - 9 - 10 1098 4460290 68
S63463.1 - 9 - 10 1170 68
X17459.1 - 9 - 10 1165 68

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbpj

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
angiogenesis
somitogenesis
epithelial to mesenchymal transition
heart looping
blood vessel remodeling
inflammatory response to antigenic stimulus
secondary heart field specification
outflow tract morphogenesis
endocardium development
endocardium morphogenesis
epithelial to mesenchymal transition involved in endocardial cushion formation
cardiac left ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
humoral immune response
Notch signaling pathway
positive regulation of transcription of Notch receptor target
heart development
positive regulation of cell proliferation
negative regulation of cell proliferation
auditory receptor cell fate commitment
epidermal cell fate specification
regulation of gene expression
positive regulation of gene expression
negative regulation of neuron projection development
negative regulation of smooth muscle cell migration
pituitary gland development
hemopoiesis
neuron differentiation
B cell differentiation
keratinocyte differentiation
negative regulation of ossification
positive regulation of BMP signaling pathway
negative regulation of smooth muscle cell apoptotic process
somatic stem cell population maintenance
positive regulation of Notch signaling pathway involved in heart induction
dorsal aorta morphogenesis
atrioventricular canal development
defense response to bacterium
myeloid dendritic cell differentiation
cell fate commitment
negative regulation of cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of timing of cell differentiation
positive regulation of smooth muscle cell proliferation
sebaceous gland development
hair follicle maturation
artery morphogenesis
positive regulation of cardiac muscle cell proliferation
Clara cell differentiation
labyrinthine layer blood vessel development
arterial endothelial cell fate commitment
Notch signaling pathway involved in arterial endothelial cell fate commitment
regulation of cell adhesion involved in heart morphogenesis
determination of heart left/right asymmetry
positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
blood vessel lumenization
interleukin-4 secretion
blood vessel endothelial cell fate specification
positive regulation of ERBB signaling pathway
positive regulation of ephrin receptor signaling pathway
positive regulation of canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
positive regulation of cell proliferation involved in heart morphogenesis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II repressing transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein complex binding
sequence-specific DNA binding
protein N-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500005037.mm.1 TC0500000541.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500005037.mm.1 NM_001080927.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500005037.mm.1 NM_001080928.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005037.mm.1 NM_001080928.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005037.mm.1 NM_001277116.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500005037.mm.1 NM_009035.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0