This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03047338_m1
See other ATOX1 GE Assays ›
Gene Symbol:
Atox1
Gene Name:
ATX1 (antioxidant protein 1) homolog 1 (yeast)
Gene Aliases:
AI256639, Atx1
Chromosome Location:
Chr.11: 55446643 - 55461138 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03047338_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009720.2 NP_033850.1 3 - 4 295 101
GenBank mRNA AF004591.1 - 3 - 4 288 101
AK002432.1 - 3 - 4 300 101
BC027632.1 - 3 - 4 295 1328907 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002784.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atox1

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
copper ion transport
cellular copper ion homeostasis
response to oxidative stress
intracellular copper ion transport
metal ion transport
copper ion transmembrane transport
copper ion transmembrane transporter activity
copper ion binding
copper chaperone activity
copper-dependent protein binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100025290.mm.1 NM_009720.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025290.mm.1 NM_009720.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100025290.mm.1 TC1100002784.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 589987 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0