This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03047554_g1
See other GTF2B GE Assays ›
Gene Symbol:
Gtf2b
Gene Name:
general transcription factor IIB
Gene Aliases:
-
Chromosome Location:
Chr.3: 142765247 - 142783606 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03047554_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145546.1 NP_663521.1 5 - 6 566 107
GenBank mRNA AK131856.1 - 5 - 6 588 107
AK133097.1 - 5 - 6 588 107
AK133825.1 - 5 - 6 588 107
AK145403.1 - 5 - 6 588 107
AK146223.1 - 5 - 6 589 107
AK146666.1 - 5 - 6 588 107
AK146910.1 - 5 - 6 588 107
AK151577.1 - 5 - 6 588 107
BC016637.1 - 5 - 6 566 2650779 107

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2b

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
core promoter binding
DNA binding
protein binding
transcription factor binding
zinc ion binding
TBP-class protein binding
metal ion binding
thyroid hormone receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300011722.mm.1 NM_145546.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300011722.mm.1 NM_145546.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300011722.mm.1 TC0300001449.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline