This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03058253_g1
See other SAT2 GE Assays ›
Gene Symbol:
Sat2
Gene Name:
spermidine/spermine N1-acetyl transferase 2
Gene Aliases:
2610016A03Rik, SSAT-2, SSAT2
Chromosome Location:
Chr.11: 69621993 - 69624034 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03058253_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026991.2 NP_081267.1 2 - 3 278 61
GenBank mRNA AK011406.1 - 3 - 4 196 61
AK161230.1 - 2 - 3 336 61
BC061227.1 - 2 - 3 278 30312881 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000917.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

spermine metabolic process
spermidine metabolic process
spermidine acetylation
spermine acetylation
putrescine acetylation
nor-spermidine metabolic process
diamine N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100007776.mm.1 NM_026991.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100007776.mm.1 NM_026991.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100007776.mm.1 TC1100000917.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0