This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03456263_s1
See other TERC GE Assays ›
Gene Symbol:
Terc
Gene Name:
telomerase RNA component
Gene Aliases:
mTER, mTR
Chromosome Location:
Chr.3: 96414437 - 96414833 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03456263_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Terc
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_001579.1 LIT2118
telomerase RNA
1 251 143
GenBank mRNA AK085092.1 -
252 143
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU923232.1 - 323 Chr. - - [-] on Build 143
GenBankmRNA BB660202.1 - 1 248 Chr. 3 - 96414207 - 96414830 [-] on Build GRCm38 143

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream U1b2 small nuclear RNA Rnu1b2 pU1.2 -40.3kb Anti-sense Chr. 3 - 96374021 - 96374185 on NCBI Build GRCm38
Downstream cDNA sequence BC107364 BC107364 D730018G16 19.0kb Sense Chr. 3 - 96433788 - 96454359 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
telomere maintenance via telomerase
cell proliferation
post-embryonic development
multicellular organism aging
positive regulation of telomere maintenance via telomerase
regulation of cell proliferation
hematopoietic stem cell differentiation
DNA biosynthetic process
template for synthesis of G-rich strand of telomere DNA activity
telomerase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544476 AK085092.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544476 NR_001579.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544476 AK085092.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P544476 NR_001579.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413040 - -
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P544476 AK085092.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P544476 NR_001579.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P544476 AK085092.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P544476 NR_001579.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P248935 AK085092.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P248935 AK085092.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P544476 AK085092.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P544476 NR_001579.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P544476 AK085092.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P544476 NR_001579.1 mouse_transcriptome
Agilent Six_ChIP_01_of_03_v2 A_68_P22349239 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349256 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349231 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02464077 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349232 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349243 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349261 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349235 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349254 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349255 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02567678 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349251 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349265 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349269 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349250 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349260 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02868572 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349253 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349241 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02636541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349234 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22349262 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10500346 - -
Affymetrix Mouse Gene 1.1 ST Array 10500345 - -
Affymetrix Mouse Clariom S Array PSR0300019593.mm.1 NR_001579.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019593.mm.1 NR_001579.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019593.mm.1 TC0300002512.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 687306 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105690450 - -
Illumina mouse-6 v1.0 expression beadchip 105690450 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2522855 - -
mouse-6 v1.1 (Illumina) scl11785.1.1_125-S - -
Illumina Sentrix mouse-6.v1 bead array 105690450 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 105690450 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2522855 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0