This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03807841_g1
See other PPM1B GE Assays ›
Gene Symbol:
Ppm1b
Gene Name:
protein phosphatase 1B, magnesium dependent, beta isoform
Gene Aliases:
PP2CB
Chromosome Location:
Chr.17: 84956741 - 85023992 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03807841_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppm1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159496.1 NP_001152968.1 2 - 3 1255 100
NM_001159497.1 NP_001152969.1 2 - 3 1008 100
NM_001159498.1 NP_001152970.1 2 - 3 931 100
NM_011151.2 NP_035281.1 2 - 3 1255 100
GenBank mRNA AJ271833.1 - 2 - 3 1233 100
AJ271836.1 - 2 - 3 1241 100
AK010224.1 - 1 - 2 158 100
AK050170.1 - 2 - 3 1256 100
AK077136.1 - 3 - 4 1003 100
AK136077.1 - 2 - 3 1257 100
AK160653.1 - 2 - 3 1256 100
AK163283.1 - 2 - 3 1248 100
AK167176.1 - 2 - 3 1254 100
BC018556.1 - 2 - 3 946 3987825 100
BC090963.1 - 1 - 2 846 30632168 100
BQ176513.1 - 5 - 6 648 98
D17411.1 - 2 - 3 880 100
D17412.1 - 2 - 3 880 100
D45859.1 - 1 - 2 127 100
D45860.1 - 2 - 3 880 100
D45861.1 - 1 - 2 127 100
U09218.1 - 2 - 3 939 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppm1b

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
N-terminal protein myristoylation
negative regulation of interferon-beta production
peptidyl-threonine dephosphorylation
negative regulation of NF-kappaB import into nucleus
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of defense response to virus
magnesium ion binding
catalytic activity
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
manganese ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline