This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929380_m1
See other IGSF3 GE Assays ›
Gene Symbol:
Igsf3
Gene Name:
immunoglobulin superfamily, member 3
Gene Aliases:
1700016K10Rik, 2810035F16Rik, 4833439O17Rik, mKIAA0466
Chromosome Location:
Chr.3: 101377011 - 101463060 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929380_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006042.1 - 1 - 2 107 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706159.1 - 1 - 2 104 Chr. 3 - 101425274 - 101426936 [-] on Build GRCm38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CD2 antigen Cd2 LFA-2,
Ly-37,
Ly37
-137.3kb Sense Chr. 3 - 101275908 - 101287939 on NCBI Build GRCm38
Downstream ATPase,
Na+/K+ transporting,
alpha 1 polypeptide
Atp1a1 Atpa-1,
BC010319
149.3kb Sense Chr. 3 - 101576219 - 101604707 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 796971 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0