This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929429_g1
See other PRSS23OS GE Assays ›
Gene Symbol:
Prss23os
Gene Name:
protease, serine 23, opposite strand
Gene Aliases:
1700019G06Rik, 4930479H08Rik
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929429_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss23os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006119.1 - 1 - 2 163 62
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706225.1 - 1 - 2 163 Chr. 7 - 89510023 - 89510490 [+] on Build GRCm38 62
GenBankmRNA CR515421.1 - 1 - 2 Chr. 7 - 89510059 - 89510493 [-] on Build GRCm38 62

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream expressed sequence AI314278 AI314278 -77.9kb Sense Chr. 7 - 89426306 - 89432077 on NCBI Build GRCm38
Downstream malic enzyme 3,
NADP(+)-dependent,
mitochondrial
Me3 1700020C08Rik,
B230207H15Rik
121.7kb Sense Chr. 7 - 89632197 - 89854359 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113599 BC018517.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113599 AK156348.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113599 AK033671.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113599 AK034439.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113599 AK078518.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113599 BC018517.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113599 AK156348.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113599 AK033671.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113599 AK034439.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113599 AK078518.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101740504 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline