This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03933073_s1
See other FIRRE GE Assays ›
Gene Symbol:
Firre
Gene Name:
functional intergenic repeating RNA element
Gene Aliases:
5830467J12Rik, 6720401G13Rik, AW048145
Chromosome Location:
Chr.X: 50555744 - 50635258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03933073_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Firre
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK032694.1 - 1 1251 89
BC055934.1 - 3 1780 89
BP762728.1 - 1 89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM069026.1 - 1 Chr. X - 50563185 - 50563571 [+] on Build GRCm38 89
GenBankmRNA BF227957.1 - 1 Chr. X - 50563122 - 50563532 [+] on Build GRCm38 89
GenBankmRNA AA920161.1 - 1 156 Chr. X - 50563223 - 50563596 [-] on Build GRCm38 89
GenBankmRNA BP760824.1 - 1 Chr. X - 50563120 - 50563617 [+] on Build GRCm38 89
GenBankmRNA AI482406.1 - 1 Chr. X - 50563185 - 50563540 [+] on Build GRCm38 89
GenBankmRNA CB205241.1 - 1 458 Chr. X - 50563142 - 50563898 [-] on Build GRCm38 89
GenBankmRNA CB196487.1 - 1 677 Chr. X - 50563287 - 50564117 [-] on Build GRCm38 89
GenBankmRNA BF227221.1 - 1 Chr. X - 50563122 - 50563551 [+] on Build GRCm38 89
GenBankmRNA AW322499.1 - 1 Chr. X - 50563125 - 50563564 [+] on Build GRCm38 89
GenBankmRNA AW556424.2 - 1 Chr. X - 50563187 - 50563584 [+] on Build GRCm38 89
GenBankmRNA BU703657.1 - 1 97 Chr. X - 50563112 - 50563536 [-] on Build GRCm38 89
GenBankmRNA BE447060.1 - 1 Chr. X - 50563185 - 50563648 [+] on Build GRCm38 89
GenBankmRNA BG075556.2 - 1 Chr. X - 50563187 - 50563728 [+] on Build GRCm38 89
GenBankmRNA BP753960.1 - 1 Chr. X - 50563127 - 50563478 [+] on Build GRCm38 89
GenBankmRNA AI552904.1 - 1 133 Chr. X - 50563185 - 50563573 [-] on Build GRCm38 89
GenBankmRNA BP771062.1 - 1 Chr. X - 50563127 - 50563700 [+] on Build GRCm38 89
GenBankmRNA BM068949.1 - 1 Chr. X - 50563185 - 50563665 [+] on Build GRCm38 89
GenBankmRNA AW048145.1 - 1 Chr. X - 50563185 - 50563620 [+] on Build GRCm38 89
GenBankmRNA AA462795.1 - 1 134 Chr. X - 50563205 - 50563568 [-] on Build GRCm38 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
35612
Gm35612 Anti-sense Chr. X - 50537345 - 50591465 on NCBI Build GRCm38
Downstream serine/threonine kinase 26 Stk26 2610018G03Rik,
Mst4
265.0kb Anti-sense Chr. X - 50829694 - 50893098 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0