This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939523_s1
See other SEC22B GE Assays ›
Gene Symbol:
Sec22b
Gene Name:
SEC22 vesicle trafficking protein homolog B (S. cerevisiae)
Gene Aliases:
4930564D15Rik, AA517334, AI480645, C81333, ERS-24, Sec22l1
Chromosome Location:
Chr.3: 97901227 - 97922318 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939523_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec22b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016219.1 - 247 91
AK132726.1 - 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DV066700.1 - 1 436 Chr. 3 - 97922522 - 97923345 [+] on Build GRCm38 91
GenBankmRNA BU056157.1 - 1 101 Chr. 3 - 97922819 - 97923276 [+] on Build GRCm38 91
GenBankmRNA BM940032.1 - 1 Chr. 3 - 97922713 - 97923276 [-] on Build GRCm38 91
GenBankmRNA BQ552557.1 - 1 Chr. 3 - 97922763 - 97923275 [-] on Build GRCm38 91
GenBankmRNA CB318283.1 - 89 Chr. - - [-] on Build 91
GenBankmRNA BE648328.1 - 1 199 Chr. 3 - 97922721 - 97923276 [+] on Build GRCm38 91
GenBankmRNA BQ257487.1 - 1 153 Chr. 3 - 97922778 - 97923276 [+] on Build GRCm38 91
GenBankmRNA C81202.1 - Chr. 4 - 85629970 - 85629985 [+] on Build GRCm38 91
GenBankmRNA AI503634.1 - 1 Chr. 3 - 97922847 - 97923276 [-] on Build GRCm38 91
GenBankmRNA BQ552558.1 - 1 382 Chr. 3 - 97922543 - 97923112 [+] on Build GRCm38 91
GenBankmRNA BG088999.1 - 1 Chr. 3 - 97922628 - 97923172 [-] on Build GRCm38 91
GenBankmRNA AA267140.1 - 1 324 Chr. 3 - 97922596 - 97923063 [+] on Build GRCm38 91
GenBankmRNA AA692530.1 - 1 63 Chr. 3 - 97922857 - 97923270 [+] on Build GRCm38 91
GenBankmRNA AW539497.1 - 1 Chr. 3 - 97922816 - 97923287 [-] on Build GRCm38 91
GenBankmRNA C81333.1 - Chr. 17 - 21921166 - 21921185 [+] on Build GRCm38 91
GenBankmRNA BY715826.1 - 1 247 Chr. 3 - 97922673 - 97923275 [+] on Build GRCm38 91
GenBankmRNA AA146117.1 - 1 63 Chr. 3 - 97922857 - 97923261 [+] on Build GRCm38 91
GenBankmRNA BQ175529.1 - 1 Chr. 3 - 97922558 - 97923276 [-] on Build GRCm38 91
GenBankmRNA CK376045.1 - 1 178 Chr. 3 - 97922763 - 97923276 [+] on Build GRCm38 91
GenBankmRNA BE307891.1 - 1 Chr. 3 - 97922393 - 97923004 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Downstream predicted gene 5544 Gm5544 EG433632 6.9kb Sense Chr. 3 - 97930173 - 97967018 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 554899 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0