This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939606_m1
See other TMIGD3 GE Assays ›
Gene Symbol:
Tmigd3
Gene Name:
transmembrane and immunoglobulin domain containing 3
Gene Aliases:
1700001d09rik, 4930578J19Rik
Chromosome Location:
Chr.3: 105870858 - 105924044 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939606_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmigd3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016304.1 - 1 - 2 109 98
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715940.1 - 1 - 2 109 Chr. 3 - 105870903 - 105872632 [+] on Build GRCm38 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 5547 Gm5547 EG433637 -53.5kb Sense Chr. 3 - 105815567 - 105817359 on NCBI Build GRCm38
Downstream adenosine A3 receptor Adora3 A3AR,
A3R,
AA3R,
ARA3,
Gpcr2
31.8kb Sense Chr. 3 - 105904421 - 105908928 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0