This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03940510_g1
See other ATP5L GE Assays ›
Gene Symbol:
Atp5l
Gene Name:
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G
Gene Aliases:
4933437C06Rik
Chromosome Location:
Chr.9: 44913248 - 44920742 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03940510_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017092.1 - 555 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus 1700

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 836234 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0