This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03942089_s1
See other MEG3 GE Assays ›
Gene Symbol:
Meg3
Gene Name:
maternally expressed 3
Gene Aliases:
2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394
Chromosome Location:
Chr.12: 109540996 - 109571729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03942089_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Meg3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003633.3 5 5528 101
NR_027651.2 9 5516 101
GenBank mRNA AK018142.1 - 1 655 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV511819.1 - 1 358 Chr. 12 - 109562349 - 109562834 [+] on Build GRCm38 101
GenBankmRNA BQ746598.1 - 1 309 Chr. 12 - 109562399 - 109563014 [+] on Build GRCm38 101
GenBankmRNA BY269973.1 - 1 394 Chr. 12 - 109562313 - 109562763 [+] on Build GRCm38 101
GenBankmRNA CB059172.1 - 1 253 Chr. 12 - 109562455 - 109563135 [+] on Build GRCm38 101
GenBankmRNA CB203164.1 - 1 388 Chr. 12 - 109562319 - 109563179 [+] on Build GRCm38 101
GenBankmRNA AI850416.1 - 1 293 Chr. 12 - 109562436 - 109562788 [+] on Build GRCm38 101
GenBankmRNA BG298422.1 - 1 344 Chr. 12 - 109562362 - 109563061 [+] on Build GRCm38 101
GenBankmRNA BF472073.1 - 1 248 Chr. 12 - 109562459 - 109563136 [+] on Build GRCm38 101
GenBankmRNA BG298969.1 - 345 Chr. - - [-] on Build 101
GenBankmRNA CX243886.1 - 1 381 Chr. 12 - 109562326 - 109563129 [+] on Build GRCm38 101
GenBankmRNA CB058829.1 - 1 316 Chr. 12 - 109562399 - 109563085 [+] on Build GRCm38 101
GenBankmRNA BQ746587.1 - 1 308 Chr. 12 - 109562399 - 109563135 [+] on Build GRCm38 101
GenBankmRNA BQ177683.1 - 1 163 Chr. 12 - 109562544 - 109563198 [+] on Build GRCm38 101
GenBankmRNA CB057805.1 - 1 191 Chr. 12 - 109562515 - 109563175 [+] on Build GRCm38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1906-1 Mir1906-1 Mir1906,
Mirn1906,
mmu-mir-1906,
mmu-mir-1906-1
-17.4kb Sense Chr. 12 - 109544541 - 109544620 on NCBI Build GRCm38
Downstream microRNA 770 Mir770 Mirn770,
mir-770,
mmu-mir-770
429.0 Sense Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

in utero embryonic development
liver development
DNA methylation
skeletal muscle tissue development
determination of adult lifespan
post-embryonic development
multicellular organism growth
lung alveolus development
SMAD protein signal transduction
genetic imprinting

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200008087.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008087.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008087.mm.1 NR_030427.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008087.mm.1 NR_030427.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008087.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008084.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008084.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008084.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008086.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008086.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008086.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008083.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008083.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008083.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008085.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008085.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008085.mm.1 TC1200001127.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline