This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03949215_s1
See other SSBP2 GE Assays ›
Gene Symbol:
Ssbp2
Gene Name:
single-stranded DNA binding protein 2
Gene Aliases:
1500004K09Rik, 2310079I02Rik, 9330163K02Rik, A830008M03Rik, AU067692, AW558684, Hspc116, SSDP2
Chromosome Location:
Chr.13: 91460979 - 91706175 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03949215_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024186.5 NP_077148.1 16 6418 95
NM_024272.4 NP_077234.2 17 6508 95
GenBank mRNA AK140460.1 - 1 1840 95
AK163666.1 - 1 1841 95
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB521645.1 - 1 187 Chr. 13 - 91705841 - 91706122 [+] on Build GRCm38 95
GenBankmRNA AI465287.1 - 1 Chr. 13 - 91705761 - 91706171 [-] on Build GRCm38 95
GenBankmRNA BB243157.1 - 1 153 Chr. 13 - 91705875 - 91706172 [+] on Build GRCm38 95
GenBankmRNA BB180519.1 - 1 130 Chr. 13 - 91705898 - 91706175 [+] on Build GRCm38 95
GenBankmRNA AA218155.1 - 1 531 Chr. 13 - 91705497 - 91706111 [+] on Build GRCm38 95
GenBankmRNA BB255936.1 - 1 131 Chr. 13 - 91705897 - 91706123 [+] on Build GRCm38 95
GenBankmRNA BB265335.1 - 1 145 Chr. 13 - 91705883 - 91706166 [+] on Build GRCm38 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 4833422C13 gene 4833422C13Rik Anti-sense Chr. 13 - 91701665 - 91741872 on NCBI Build GRCm38
Downstream acyl-CoA thioesterase 12 Acot12 1300004O04Rik,
4930449F15Rik,
AV027244,
Cach,
mCACH-1
35.4kb Sense Chr. 13 - 91741511 - 91786148 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssbp2

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
DNA binding
single-stranded DNA binding
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006536.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006536.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006536.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006536.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006536.mm.1 TC1300001020.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006540.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006540.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006540.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006540.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006540.mm.1 TC1300001020.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006542.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006542.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006542.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006542.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006542.mm.1 TC1300001020.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006539.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006539.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006539.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006539.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006539.mm.1 TC1300001020.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006541.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006541.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006541.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006541.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006541.mm.1 TC1300001020.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006537.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006537.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006537.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006537.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006537.mm.1 TC1300001020.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006538.mm.1 NM_024186.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006538.mm.1 NM_024186.5 Refseq
Affymetrix Mouse Clariom S Array PSR1300006538.mm.1 NM_024272.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006538.mm.1 NM_024272.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300006538.mm.1 TC1300001020.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 486848 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0