This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03949677_m1
See other GM6300 GE Assays ›
Gene Symbol:
Gm6300
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member pseudogene
Gene Aliases:
EG622229
Chromosome Location:
Chr.3: 14363117 - 14374239 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03949677_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm6300
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033591.2 2 - 3 888 72
GenBank mRNA AK143764.1 - 2 - 3 891 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RALY RNA binding protein-like Ralyl 0710005M24Rik -188.5kb Sense Chr. 3 - 13471655 - 14182287 on NCBI Build GRCm38
Downstream solute carrier family 7 (cationic amino acid transporter,
y+ system),
member 12
Slc7a12 Asc-2,
XAT1
106.5kb Sense Chr. 3 - 14480691 - 14505819 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000088.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537515 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829466 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8220 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04831435 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300000324.mm.1 NR_033591.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300000324.mm.1 TC0300000088.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300000323.mm.1 NR_033591.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300000323.mm.1 TC0300000088.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300000322.mm.1 NR_033591.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300000322.mm.1 TC0300000088.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline