This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03951247_m1
See other SLCO2A1 GE Assays ›
Gene Symbol:
Slco2a1
Gene Name:
solute carrier organic anion transporter family, member 2a1
Gene Aliases:
2310021C19Rik, Pgt, Slc21a2, mPgt
Chromosome Location:
Chr.9: 103007668 - 103087849 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03951247_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco2a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK165716.1 - 178 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB869693.1 - 1 - 2 177 Chr. 9 - 102988712 - 103004870 [+] on Build GRCm38 111
GenBankmRNA BB870324.1 - 1 - 2 177 Chr. 9 - 102988712 - 103004868 [+] on Build GRCm38 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream receptor-like tyrosine kinase Ryk AW536699,
ERK-3,
Vik
-80.4kb Sense Chr. 9 - 102834920 - 102908307 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0