This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953633_s1
See other MSANTD2 GE Assays ›
Gene Symbol:
Msantd2
Gene Name:
Myb/SANT-like DNA-binding domain containing 2
Gene Aliases:
2810450G17Rik, 9530092B10Rik
Chromosome Location:
Chr.9: 37487773 - 37526113 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953633_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msantd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013318.1 - 1 343 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ895664.1 - 113 Chr. - - [-] on Build 90
GenBankmRNA BX519184.1 - 1 Chr. 9 - 37515845 - 37516367 [-] on Build GRCm38 90
GenBankmRNA BI180508.1 - 1 Chr. 9 - 37515795 - 37516355 [-] on Build GRCm38 90
GenBankmRNA BE334131.1 - 1 Chr. 9 - 37515795 - 37516355 [-] on Build GRCm38 90
GenBankmRNA CK639586.1 - 1 449 Chr. 9 - 37515446 - 37516142 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream roundabout homolog 3 (Drosophila) Robo3 Rbig1,
Rig-1,
Rig1,
Robo3a,
Robo3b
-82.1kb Anti-sense Chr. 9 - 37416045 - 37433408 on NCBI Build GRCm38
Downstream endothelial cell-specific adhesion molecule Esam 2310008D05Rik,
Esam1,
W117m
11.7kb Sense Chr. 9 - 37528055 - 37538319 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 370730 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0