This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953951_s1
See other NAA20 GE Assays ›
Gene Symbol:
Naa20
Gene Name:
N(alpha)-acetyltransferase 20, NatB catalytic subunit
Gene Aliases:
1500004D14Rik, 2900026I01Rik, AU041458, D2Ertd186e, Nat5
Chromosome Location:
Chr.2: 145903206 - 145916425 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953951_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013601.1 - 1 355 61
AK046153.1 - 1 61
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN709072.1 - 1 Chr. 2 - 145910188 - 145910808 [+] on Build GRCm38 61
GenBankmRNA BY712563.1 - 1 352 Chr. 2 - 145910647 - 145911071 [-] on Build GRCm38 61
GenBankmRNA AV151985.1 - 1 203 Chr. 2 - 145910648 - 145910914 [-] on Build GRCm38 61

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
39954
Gm39954 Sense Chr. 2 - 145880114 - 145915869 on NCBI Build GRCm38
Downstream Crn,
crooked neck-like 1 (Drosophila)
Crnkl1 1200013P10Rik,
5730590A01Rik,
C80326,
crn
6.4kb Sense Chr. 2 - 145917482 - 145934700 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 495618 - -
ABI Mouse Genome Survey Microarray 749027 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0