Assay Details
| Gene Symbol: 4833403J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014653.1 | - | 1 | 349 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
GTPase, IMAP family member 5 |
Gimap5 |
D630024P16, E230026N22Rik |
-103.0 | Sense | Chr. 6 - 48746137 - 48755235 on NCBI Build GRCm38 |
| Downstream |
GTPase, IMAP family member 3 |
Gimap3 |
2010110D23Rik, Gimap5, Ian4 |
8.1kb | Anti-sense | Chr. 6 - 48764464 - 48770851 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4833403J16Rik
Genomic Map